Learn More
Medchemexpress LLC Glycerol phenylbutyrate | 611168-24-2 | 99.8% | 1 ML
SDP
Catalog No. 5000411483
Encompass_Preferred
Glycerol phenylbutyrate is a sigma-2 (σ2) receptor ligand and a new generation ammonia scavenger agent. It has potential for the treatment of hyperammonemia and may also have therapeutic potential in conditions such as chronic hepatic encephalopathy or other inherited metabolic disorders.
  • Sigma-2 (σ2) receptor ligand.
  • New generation ammonia scavenger agent.
  • Potential for treatment of hyperammonemia.
  • May have therapeutic potential in chronic hepatic encephalopathy or other inherited metabolic disorders.
Catalog No. 50-004-11483 Supplier Medchemexpress LLC Supplier No. HYB208710MM/1ML
Glycerol phenylbutyrate is a sigma-2 (σ2) receptor ligand and a new generation ammonia scavenger agent. It has potential for the treatment of hyperammonemia and may also have therapeutic potential in conditions such as chronic hepatic encephalopathy or other inherited metabolic disorders.
  • Sigma-2 (σ2) receptor ligand.
  • New generation ammonia scavenger agent.
  • Potential for treatment of hyperammonemia.
  • May have therapeutic potential in chronic hepatic encephalopathy or other inherited metabolic disorders.