Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycerol phenylbutyrate | 611168-24-2 | 99.8% | 100 MG
SDP

Catalog No. 5000410743
Encompass_Preferred

Glycerol phenylbutyrate is identified as a sigma-2 (σ2) receptor ligand with a pKi of 8.02. It functions as a new generation ammonia scavenger agent. This compound is intended for research use only and is not sold to patients. It has potential for treating hyperammonemia and may also be therapeutic for conditions such as chronic hepatic encephalopathy or other inherited metabolic disorders.

  • Purity: 99.81%
  • Available in various sizes, including 10 mM * 1 mL in DMSO (solution), 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, and 500 mg.
  • Solubility: 100 mg/mL in DMSO, 50 mg/mL in ethanol, and < 0.1 mg/mL in H2O.
  • Storage conditions for pure form: -20°C for 3 years, 4°C for 2 years.
  • Storage conditions for solution in solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-004-10743 Supplier Medchemexpress LLC Supplier No. HYB2087100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycerol phenylbutyrate is identified as a sigma-2 (σ2) receptor ligand with a pKi of 8.02. It functions as a new generation ammonia scavenger agent. This compound is intended for research use only and is not sold to patients. It has potential for treating hyperammonemia and may also be therapeutic for conditions such as chronic hepatic encephalopathy or other inherited metabolic disorders.

  • Purity: 99.81%
  • Available in various sizes, including 10 mM * 1 mL in DMSO (solution), 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, and 500 mg.
  • Solubility: 100 mg/mL in DMSO, 50 mg/mL in ethanol, and < 0.1 mg/mL in H2O.
  • Storage conditions for pure form: -20°C for 3 years, 4°C for 2 years.
  • Storage conditions for solution in solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.