Learn More
Medchemexpress LLC Glycerol phenylbutyrate | 611168-24-2 | 99.8% | 5 MG
SDP
Catalog No. 5000411482
Encompass_Preferred
Glycerol phenylbutyrate, also known as HPN-100, is a sigma-2 (σ2) receptor ligand with a pKi of 8.02. It serves as a new generation ammonia scavenger agent and is intended for research use only. This compound has the potential for treating hyperammonemia and may be therapeutically beneficial for conditions such as chronic hepatic encephalopathy or other inherited metabolic disorders.
  • Acts as a sigma-2 (σ2) receptor ligand
  • New generation ammonia scavenger agent
  • Potential treatment for hyperammonemia
  • May benefit chronic hepatic encephalopathy
  • May benefit inherited metabolic disorders
Catalog No. 50-004-11482 Supplier Medchemexpress LLC Supplier No. HYB20875MG
Glycerol phenylbutyrate, also known as HPN-100, is a sigma-2 (σ2) receptor ligand with a pKi of 8.02. It serves as a new generation ammonia scavenger agent and is intended for research use only. This compound has the potential for treating hyperammonemia and may be therapeutically beneficial for conditions such as chronic hepatic encephalopathy or other inherited metabolic disorders.
  • Acts as a sigma-2 (σ2) receptor ligand
  • New generation ammonia scavenger agent
  • Potential treatment for hyperammonemia
  • May benefit chronic hepatic encephalopathy
  • May benefit inherited metabolic disorders