Learn More
Selleck Chemical LLC Glycerol phenylbutyrate
SDP
Catalog No. 5000002757
Encompass_Preferred
Glycerol phenylbutyrate (GT4P HPN-100M Ravicti) is a nitrogen-binding agent and acts as a sigma-2 ( 2) receptor ligand with a pKi of 8 02 Glycerol phenylbutyrate is a prodrug in which phenylbutyrate (PBA) is released from the glycerol backbone by lipases in the gastrointestinal tract
Catalog No. 50-000-02757 Supplier Selleck Chemical LLC Supplier No. S69815MG
Glycerol phenylbutyrate (GT4P HPN-100M Ravicti) is a nitrogen-binding agent and acts as a sigma-2 ( 2) receptor ligand with a pKi of 8 02 Glycerol phenylbutyrate is a prodrug in which phenylbutyrate (PBA) is released from the glycerol backbone by lipases in the gastrointestinal tract