Learn More
Sigma Aldrich Fine Chemicals Biosciences Glycerol puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E422, anhydrous, 99.0-101.0% (alkalimetric) | 56-81-5 | MFCD00004722 | 1l
Catalog No. 501754374
Encompass
Glycerol puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E422, anhydrous, 99.0-101.0% (alkalimetric) | Mol Wt: 92.09 | 56-81-5 | MFCD00004722 | 1l
Catalog No. 50-175-4374 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 155231LR
Glycerol puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E422, anhydrous, 99.0-101.0% (alkalimetric) | Mol Wt: 92.09 | 56-81-5 | MFCD00004722 | 1l