Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol tripalmitate, 98%

Catalog No. AAA1092206
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1092206 5 g
AAA1092203 1 g
AAA1092214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1092206 Supplier Thermo Scientific Chemicals Supplier No. A1092206
Only null left
Add to Cart

CAS: 555-44-2 | C51H98O6 | 807.339 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-44-2
Molecular Formula C51H98O6
Molecular Weight (g/mol) 807.339
MDL Number MFCD00008995
InChI Key PVNIQBQSYATKKL-UHFFFAOYSA-N
Synonym tripalmitin, glycerol tripalmitate, tripalmitoylglycerol, glyceryl tripalmitate, palmitic triglyceride, tripalmitate, triglyceryl palmitate, barolub, dynasan 116, propane-1,2,3-triyl tripalmitate
PubChem CID 11147
ChEBI CHEBI:77393
IUPAC Name 2,3-di(hexadecanoyloxy)propyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC

Specifications

Melting Point 60°C to 64°C
Quantity 5 g
Formula Weight 807.34
Percent Purity 98%
Chemical Name or Material Glycerol tripalmitate
EINECSNumber 209-098-1
RTECSNumber RT4953500
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.