Learn More
Medchemexpress LLC Glycerol tristearate-d105 | 125941-88-0 | 99.6% | 997.13 g·mol⁻¹ | C57H5D105O6 | 5 MG
SDP
Catalog No. 5000263957
Encompass_Preferred
Tristearin-d105 is the deuterium-labeled form of tristearin, a triglyceride composed of three stearic acid units. Supplied as a high-purity, isotope-labeled standard, it is intended for research use as a tracer or internal standard in quantitative analyses and for formulation studies involving lipid-based delivery systems.
  • Deuterium-labeled triglyceride for tracing studies.
  • High purity suitable for quantitative NMR, GC-MS, and LC-MS.
  • Suitable as an internal standard for mass spectrometry assays.
  • Applicable to solid-lipid nanoparticle formulation research.
  • Supplied in small, weighed quantities optimized for analytical workflows.
Catalog No. 50-002-63957 Supplier Medchemexpress LLC Supplier No. HY127035S5MG
Tristearin-d105 is the deuterium-labeled form of tristearin, a triglyceride composed of three stearic acid units. Supplied as a high-purity, isotope-labeled standard, it is intended for research use as a tracer or internal standard in quantitative analyses and for formulation studies involving lipid-based delivery systems.
  • Deuterium-labeled triglyceride for tracing studies.
  • High purity suitable for quantitative NMR, GC-MS, and LC-MS.
  • Suitable as an internal standard for mass spectrometry assays.
  • Applicable to solid-lipid nanoparticle formulation research.
  • Supplied in small, weighed quantities optimized for analytical workflows.