Learn More
Alkali Scientific Glycerol, USP Grade, 4 L
SDP
Catalog No. 5000660074
Encompass_Preferred
Glycerol, USP Grade, 4 L, is a highly purified laboratory-grade humectant widely used in chemical and biochemical research. It is essential in maintaining moisture balance in various products, including cosmetics, pharmaceuticals, and biological solutions. In research, glycerol is used in cryopreservation to safeguard cells and tissues against ice damage during freezing. It is also used in buffer solutions, enhancing the stability and functionality of proteins and enzymes. As a humectant, it attracts and retains moisture, preventing dryness and degradation in formulations. This product meets the USP standards for purity, ensuring its reliability in sensitive laboratory and industrial applications.
Catalog No. 50-006-60074 Supplier Alkali Scientific Supplier No. LYC003CP
Glycerol, USP Grade, 4 L, is a highly purified laboratory-grade humectant widely used in chemical and biochemical research. It is essential in maintaining moisture balance in various products, including cosmetics, pharmaceuticals, and biological solutions. In research, glycerol is used in cryopreservation to safeguard cells and tissues against ice damage during freezing. It is also used in buffer solutions, enhancing the stability and functionality of proteins and enzymes. As a humectant, it attracts and retains moisture, preventing dryness and degradation in formulations. This product meets the USP standards for purity, ensuring its reliability in sensitive laboratory and industrial applications.