Learn More
Medchemexpress LLC Glycerophospho-N-palmitoyl ethanolamine | 100575-09-5 | 99.0% | 453.55 | 1 MG
SDP
Catalog No. 5000437106
Encompass_Preferred
Glycerophospho-N-palmitoyl ethanolamine (GP-NPEA) is a metabolic precursor of palmitoyl ethanolamide PEA. It decreases in the cortex of CUMS rats, which may be related to a disorder in the endocannabinoid system arising after the onset of depression.
  • Metabolic precursor of palmitoyl ethanolamide PEA.
  • Decreases in the cortex of CUMS rats.
  • May be related to a disorder in the endocannabinoid system.
Catalog No. 50-004-37106 Supplier Medchemexpress LLC Supplier No. HY1125281MG
Glycerophospho-N-palmitoyl ethanolamine (GP-NPEA) is a metabolic precursor of palmitoyl ethanolamide PEA. It decreases in the cortex of CUMS rats, which may be related to a disorder in the endocannabinoid system arising after the onset of depression.
  • Metabolic precursor of palmitoyl ethanolamide PEA.
  • Decreases in the cortex of CUMS rats.
  • May be related to a disorder in the endocannabinoid system.