Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycerophospho-N-palmitoyl ethanolamine | 100575-09-5 | MFCD11977724 | 99.0% | 453.55 | C21H44NO7P | 5 MG
SDP

Catalog No. 5000262979
Encompass_Preferred

Glycerophospho-N-palmitoyl ethanolamine is an endogenous glycerophospho lipid supplied as a white to off-white solid for research use. It is intended for use as a biochemical reagent or reference standard in studies of lipid metabolism and signaling and is supplied with supporting documentation including a product data sheet and safety data sheet.

  • Endogenous glycerophospho lipid suitable for research use.
  • Appearance: white to off-white solid.
  • Purity: 99.0%.
  • Chemical formula: C21H44NO7P; molecular weight: 453.55 g·mol⁻¹.
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months).
  • Provided with product data sheet and safety data sheet.

Catalog No. 50-002-62979 Supplier Medchemexpress LLC Supplier No. HY1125285MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycerophospho-N-palmitoyl ethanolamine is an endogenous glycerophospho lipid supplied as a white to off-white solid for research use. It is intended for use as a biochemical reagent or reference standard in studies of lipid metabolism and signaling and is supplied with supporting documentation including a product data sheet and safety data sheet.

  • Endogenous glycerophospho lipid suitable for research use.
  • Appearance: white to off-white solid.
  • Purity: 99.0%.
  • Chemical formula: C21H44NO7P; molecular weight: 453.55 g·mol⁻¹.
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months).
  • Provided with product data sheet and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.