Learn More
Cayman Chemical GlycerophosphoNOleoyl Ethanol
SDP
Supplier:  Cayman Chemical 100113571 MG
Encompass_Preferred
The precursor of OEA, an endogenous, potent agonist for PPARα (EC50 = 120 nM); OEA suppresses food intake and reduces weight gain in rats and PPARα wild-type mice, but not in PPARα knockout mice
Catalog No. 50-289-1277