Learn More
Cayman Chemical Glycerophospho-N-Oleoyl Ethanolamine, Purity: ≥98%, CAS: 201738-24-1, 10 mg
SDP
Catalog No. 502890088
Encompass_Preferred
The precursor of OEA, an endogenous, potent agonist for PPARα (EC50 = 120 nM); OEA suppresses food intake and reduces weight gain in rats and PPARα wild-type mice, but not in PPARα knockout mice
Catalog No. 50-289-0088 Supplier Cayman Chemical Supplier No. 1001135710 MG
The precursor of OEA, an endogenous, potent agonist for PPARα (EC50 = 120 nM); OEA suppresses food intake and reduces weight gain in rats and PPARα wild-type mice, but not in PPARα knockout mice