Learn More
Cayman Chemical GlycerophosphoNPalmitoyl Etha
SDP
Supplier:  Cayman Chemical 100113561 MG
Encompass_Preferred
Precursor of PEA, an endogenous CB found in brain, liver, and other mammalian tissues; PEA has potent anti-inflammatory activity in vivo
Catalog No. 50-289-1400