Learn More
Cayman Chemical Glycerophospho-N-Palmitoyl Ethanolamine, Purity: ≥95%, CAS: 100575-09-5, 5 mg
SDP
Catalog No. 502890388
Encompass_Preferred
Precursor of PEA, an endogenous CB found in brain, liver, and other mammalian tissues; PEA has potent anti-inflammatory activity in vivo
Catalog No. 50-289-0388 Supplier Cayman Chemical Supplier No. 100113565 MG
Precursor of PEA, an endogenous CB found in brain, liver, and other mammalian tissues; PEA has potent anti-inflammatory activity in vivo