Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glyceryl 1,3-dimyristate | 7770-09-4 | MFCD00048455 | 98.0% | 512.81 g·mol⁻¹ | C31H60O5 | 250 MG
SDP

Catalog No. 5000283592
Encompass_Preferred

1,3-Dimyristoyl-glycerol (glyceryl 1,3-dimyristate) is a neutral diacylglycerol used as a biochemical reagent in studies of lipid signaling and membrane biophysics. Supplied as a white to off-white solid with high purity, it is suitable for biochemical assays and model membrane experiments.

  • Used in studies of lipid signaling and membrane biophysics.
  • Purity 98.0%.
  • Molecular formula C31H60O5.
  • Molecular weight 512.81 g·mol⁻¹.
  • Appearance white to off-white solid.
  • Recommended storage: powder at -20 °C; in solvent store at -80 °C for long term.
  • Available in research-scale quantities such as 250 MG.

Catalog No. 50-002-83592 Supplier Medchemexpress LLC Supplier No. HYW127588250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Dimyristoyl-glycerol (glyceryl 1,3-dimyristate) is a neutral diacylglycerol used as a biochemical reagent in studies of lipid signaling and membrane biophysics. Supplied as a white to off-white solid with high purity, it is suitable for biochemical assays and model membrane experiments.

  • Used in studies of lipid signaling and membrane biophysics.
  • Purity 98.0%.
  • Molecular formula C31H60O5.
  • Molecular weight 512.81 g·mol⁻¹.
  • Appearance white to off-white solid.
  • Recommended storage: powder at -20 °C; in solvent store at -80 °C for long term.
  • Available in research-scale quantities such as 250 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.