Learn More
Medchemexpress LLC Glyceryl 1,3-dipalmitate-d6 | 2692624-34-1 | 98.0% | 504.70 g/mol | C35H6D62O5 | 5 MG
SDP
Catalog No. 5000262900
Encompass_Preferred
1,3-Dipalmitin-d6 (glyceryl 1,3-dipalmitate-d6) is a deuterium-labeled isotopologue of 1,3-dipalmitin used as a stable isotope standard or tracer in analytical and research workflows. It is provided as a purified synthetic standard for laboratory research applications.
  • Deuterium-labeled standard for isotope dilution and tracer studies.
  • Suitable for mass spectrometry and lipid analysis.
  • Reported purity 98.0%.
  • Available in small quantities for analytical workflows (1 mg and 5 mg).
  • CAS number 2692624-34-1 for unambiguous identification.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-62900 Supplier Medchemexpress LLC Supplier No. HY128792S5MG
1,3-Dipalmitin-d6 (glyceryl 1,3-dipalmitate-d6) is a deuterium-labeled isotopologue of 1,3-dipalmitin used as a stable isotope standard or tracer in analytical and research workflows. It is provided as a purified synthetic standard for laboratory research applications.
  • Deuterium-labeled standard for isotope dilution and tracer studies.
  • Suitable for mass spectrometry and lipid analysis.
  • Reported purity 98.0%.
  • Available in small quantities for analytical workflows (1 mg and 5 mg).
  • CAS number 2692624-34-1 for unambiguous identification.
  • For research use only; not for human or clinical use.