Learn More
Medchemexpress LLC HY-135117 100mg Medchemexpress, Glyceryl monocaprate CAS:26402-22-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY135117100MG
Encompass_Preferred
Medchemexpress, HY-135117 100mg Glyceryl monocaprate CAS:26402-22-2 Glyceryl monocaprate (Monolaurin) is a 1-monoglyceride of capric acid against gram-positive bacterial infections. Glyceryl monocaprate (Monolaurin) has inhibitory effect on Herpes Simplex Virus (HSV) and offers an effective treatment for herpes labialiss. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9861