Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glyceryl monooleate, 36.9%, MP Biomedicals™

Catalog No. ICN10488891 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10488891 1 g
ICN10488850 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10488891 Supplier MP Biomedicals, Inc Supplier No. 0210488891
Add to Cart
Add to Cart

Chemical Identifiers

CAS 111-03-5
Molecular Formula C21H40O4
Molecular Weight (g/mol) 356.547
InChI Key RZRNAYUHWVFMIP-KTKRTIGZSA-N
Synonym monoolein, glyceryl monooleate, 2,3-dihydroxypropyl oleate, 1-monoolein, 1-oleoylglycerol, 1-oleoyl-rac-glycerol, glyceryl oleate, 1-glyceryl oleate, 1-monooleoylglycerol, glycerol 1-monooleate
PubChem CID 5283468
ChEBI CHEBI:75342
IUPAC Name 2,3-dihydroxypropyl (Z)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OCC(CO)O

Specifications

Assay Percent Range 37%
Quantity 1 g
Formula Weight 356.5
Physical Form Liquid
Chemical Name or Material Glyceryl monooleate
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.