Learn More
Medchemexpress LLC Glyceryl monostearate | 31566-31-1 | C21H42O4 | 1 ML
SDP
Catalog No. 5000374800
Encompass_Preferred
Glyceryl monostearate (GMS) is a single-tailed lipidic monoglyceride. It can be used to synthesize nanoliposomes for drug delivery applications. This compound targets liposomes and is involved in the metabolic enzyme/protease pathway.
  • Used for synthesizing nanoliposomes
  • Suitable for drug delivery research
  • Acts as a monoglyceride
  • Research use only
Catalog No. 50-003-74800 Supplier Medchemexpress LLC Supplier No. HY15122610MM/1ML
Glyceryl monostearate (GMS) is a single-tailed lipidic monoglyceride. It can be used to synthesize nanoliposomes for drug delivery applications. This compound targets liposomes and is involved in the metabolic enzyme/protease pathway.
  • Used for synthesizing nanoliposomes
  • Suitable for drug delivery research
  • Acts as a monoglyceride
  • Research use only