Learn More
Medchemexpress LLC Glyceryl triarachidate | 620-64-4 | MFCD00042879 | 99.0% | 975.64 g/mol | C63H122O6 | 100MG
SDP
Catalog No. 5000238926
Encompass_Preferred
Triarachidin (glycerol triarachidate) is an endogenous triacylglycerol used as a biochemical reagent and reference standard in lipid and metabolic research. It is a triglyceride of eicosanoic acid with molecular formula C63H122O6 and molecular weight 975.64 g/mol (CAS 620-64-4). The compound is supplied as a purified solid (99.0% purity) in multiple package sizes for laboratory use.
  • Endogenous triacylglycerol relevant to lipid metabolism studies.
  • High purity suitable for analytical and biochemical assays.
  • Solid form for easy storage and handling.
  • Well characterized molecular formula and molecular weight for calculations.
  • Available in multiple pack sizes to accommodate experimental needs.
Catalog No. 50-002-38926 Supplier Medchemexpress LLC Supplier No. HY125671100MG
Triarachidin (glycerol triarachidate) is an endogenous triacylglycerol used as a biochemical reagent and reference standard in lipid and metabolic research. It is a triglyceride of eicosanoic acid with molecular formula C63H122O6 and molecular weight 975.64 g/mol (CAS 620-64-4). The compound is supplied as a purified solid (99.0% purity) in multiple package sizes for laboratory use.
  • Endogenous triacylglycerol relevant to lipid metabolism studies.
  • High purity suitable for analytical and biochemical assays.
  • Solid form for easy storage and handling.
  • Well characterized molecular formula and molecular weight for calculations.
  • Available in multiple pack sizes to accommodate experimental needs.