Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glyceryl trioleate, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012801 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2801 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2801 Supplier MilliporeSigma Supelco Supplier No. 41679100MG
Add to Cart
Add to Cart
Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 122-32-7
Density 0.91 g/mL
Flash Point 330°C
Physical Form Neat
Linear Formula (C17H33 COOCH2)2 CHOCOC17H33
MDL Number MFCD00137563
Boiling Point 235°C to 240°C (18 mmHg)
Synonym (9 Z)9-Octadecenoic acid 1,2,3-propanetriyl ester; 1,2,3-Tri(cis-9-octadecenoyl)glycerol; Glycerol trioleate; Glycerol triolein; Oleic acid triglyceride; Oleic triglyceride; TG(18:1(9 Z)/18:1(9 Z)/18:1(9 Z)); Triolein
Type Analytical Standard
Quantity 100 mg
Formula Weight 885.43
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.