Learn More
Crescent Chemical Co Inc GLYCID ETHER 100
SDP
Supplier:  Crescent Chemical Co Inc SE21045.01
Encompass_Preferred
Glycid ether 100 for electron microscopy Mixture of aliphatic di- and triepoxides. Epoxy resin of low viscosity (ca. 100-200 mPas at 25 C) (1). Combination with ARALDITE (2). With D.E.R. 736 (3). Size - 100ML Storage Conditions - +15 C TO +30 C Catalog Number - 2104
Catalog No. 50-247-571