Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Glycidol

Catalog No. 502897557
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-289-7557 500 g
50-184-4848 100 g
50-184-4849 2.5 kg
50-184-4851 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 50-289-7557 Supplier Sigma Aldrich Supplier No. G5809500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Glycidol has been used in the synthesis of hyperbranched polyglycerol (hpg) via ring opening reaction
  • These hpgs are being considered as alternatives to pegs for developing antifouling surfaces due to their thermal and oxidative stability
  • It can also be used in the synthesis of ethoxy ethyl glycidyl ether
  • Glycidol may also be used in the preparation of chelating polymers for borate removal
TRUSTED_SUSTAINABILITY

Specifications

CAS 556-52-5
Density 1.117 g/mL (at 25°C (literature))
Boiling Point 61°C to 62°C (15 mmHg)
Molecular Formula C3H6O2
Linear Formula C3H6O2
Refractive Index n20/D 1.433 (literature)
Quantity 500 g
MDL Number MFCD00005147
Synonym (+/-)-Glycidol; (+/-)-Oxirane-2-methanol; 2,3-Epoxy-1-propanol; Glycerolglycide
Molecular Weight (g/mol) 74.08
Percent Purity 96%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.