Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Glycidol, ≥95.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019855 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9855 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9855 Supplier MilliporeSigma Supelco Supplier No. 74595100MG
Only null left
Add to Cart
Add to Cart

Glycidol is an aliphatic epoxide which may be used as a stabilizer in the manufacture of vinyl polymers, as an additive for oil and synthetic hydraulic fluids, and also as a diluent in some epoxy resins.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 780°F
CAS 556-52-5
Vapor Pressure 0.9 mmHg (25°C)
Vapor Density 2.15 (vs. air)
Density 1.117 g/mL (at 25°C)
Flash Point 72°C
Physical Form Neat
Linear Formula C3H6O2
MDL Number MFCD00005147
Boiling Point 61°C to 62°C (15 mmHg)
Synonym (+/-)-Oxirane-2-methanol; 2,3-Epoxy-1-propanol; Glycerolglycide
Type Analytical Standard
Quantity 100 mg
Formula Weight 74.08
Percent Purity ≥95.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.