Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycidyl 2-methoxyphenyl ether, 98%, Thermo Scientific™

Catalog No. AAH6177706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6177706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6177706 Supplier Thermo Scientific Chemicals Supplier No. H6177706

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2210-74-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00190158
InChI Key RJNVSQLNEALZLC-UHFFFAOYSA-N
Synonym 1-2-methoxyphenoxy-2,3-epoxypropane, guaiacol glycidyl ether, 2-2-methoxyphenoxy methyl oxirane, guajacol-glycidaether, 1,2-epoxy-3-o-methoxyphenoxy propane, glycidyl 2-methoxyphenyl ether, 1,2-epoxy-3-2-methoxyphenoxy propane, oxirane, 2-methoxyphenoxy methyl, guajacol-glycidaether german
PubChem CID 98357
IUPAC Name 2-[(2-methoxyphenoxy)methyl]oxirane
SMILES COC1=CC=CC=C1OCC2CO2

Specifications

Melting Point 41°C to 48°C
Color White
Boiling Point 152°C to 158°C
Quantity 5 g
Formula Weight 180.2
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Glycidyl 2-methoxyphenyl ether
Hazard Category H315-H317-H319
Hazard Statement GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c
EINECSNumber 218-644-8
RTECSNumber TZ3550000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.