Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycidyl 2-Methoxyphenyl Ether 98.0+%, TCI America™
SDP

Catalog No. G04165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G04165G 5 g
G041625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G04165G Supplier TCI America Supplier No. G04165G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2210-74-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00190158
InChI Key RJNVSQLNEALZLC-UHFFFAOYSA-N
Synonym 1-2-methoxyphenoxy-2,3-epoxypropane, guaiacol glycidyl ether, 2-2-methoxyphenoxy methyl oxirane, guajacol-glycidaether, 1,2-epoxy-3-o-methoxyphenoxy propane, glycidyl 2-methoxyphenyl ether, 1,2-epoxy-3-2-methoxyphenoxy propane, oxirane, 2-methoxyphenoxy methyl, guajacol-glycidaether german
PubChem CID 98357
IUPAC Name 2-[(2-methoxyphenoxy)methyl]oxirane
SMILES COC1=CC=CC=C1OCC2CO2

Specifications

Melting Point 39°C
Color White
Boiling Point 120°C
Quantity 5 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Glycidyl 2-Methoxyphenyl Ether
RTECSNumber TZ3550000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.