Learn More
Medchemexpress LLC Glycidyl linoleate | 24305-63-3 | 336.5 g/mol | C21H36O3 | 5 MG
SDP
Catalog No. 5000264134
Encompass_Preferred
Glycidyl linoleate is the glycidyl ester of linoleic acid (CAS 24305-63-3). It is an esterified lipid containing an epoxide moiety used as a small-molecule reagent and analytical standard in lipid chemistry, method development, and synthetic studies. Chemical formula C21H36O3; molecular weight 336.5 g/mol.
  • Epoxide functional group enables derivatization and ring-opening reactions.
  • Useful as an analytical standard for lipid and fatty-acid analysis.
  • Suitable for synthetic or model reaction studies involving unsaturated fatty esters.
  • Soluble in common organic solvents, facilitating chromatographic methods.
  • Supplied as a purified research reagent for laboratory use.
Catalog No. 50-002-64134 Supplier Medchemexpress LLC Supplier No. HY1314245MG
Glycidyl linoleate is the glycidyl ester of linoleic acid (CAS 24305-63-3). It is an esterified lipid containing an epoxide moiety used as a small-molecule reagent and analytical standard in lipid chemistry, method development, and synthetic studies. Chemical formula C21H36O3; molecular weight 336.5 g/mol.
  • Epoxide functional group enables derivatization and ring-opening reactions.
  • Useful as an analytical standard for lipid and fatty-acid analysis.
  • Suitable for synthetic or model reaction studies involving unsaturated fatty esters.
  • Soluble in common organic solvents, facilitating chromatographic methods.
  • Supplied as a purified research reagent for laboratory use.