Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Glycidyl methacrylate

Catalog No. 502898030
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-289-8030 500 mL
50-184-4867 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-289-8030 Supplier Sigma Aldrich Supplier No. 779342500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Glycidyl methacrylate (gma) is an epoxy-functional monomer generally used to derivatize polymers
  • It can also undergo polymerization to form poly (glycidyl methacrylate)
TRUSTED_SUSTAINABILITY

Specifications

CAS 106-91-2
Density 1.075 g/mL at 20°C, 1.042 g/mL (at 25°C (literature))
Boiling Point 189°C (lit.)
Molecular Formula C7H10O3
Refractive Index n20/D 1.449 (literature); n20/D 1.450
Linear Formula C7H10O3
MDL Number MFCD00005137
Quantity 500 mL
Synonym GMA; 2,3-Epoxypropyl methacrylate; Methacrylic acid 2,3-epoxypropyl ester
Molecular Weight (g/mol) 142.15
Percent Purity ≥97.0% (GC)
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.