Learn More
Medchemexpress LLC Glycidyl oleate-d5 | 1426395-63-2 | C21H33D5O3 | 5 MG
SDP
Catalog No. 5000372289
Encompass_Preferred
Glycidyl oleate-d5 is a deuterium labeled form of Glycidyl oleate. This compound incorporates stable heavy isotopes of hydrogen, carbon, and other elements, making it suitable for use as a tracer and internal standard in quantitative analysis. Deuteration has shown potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes for quantitation during drug development.
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-72289 Supplier Medchemexpress LLC Supplier No. HY151868S5MG
Glycidyl oleate-d5 is a deuterium labeled form of Glycidyl oleate. This compound incorporates stable heavy isotopes of hydrogen, carbon, and other elements, making it suitable for use as a tracer and internal standard in quantitative analysis. Deuteration has shown potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes for quantitation during drug development.
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs.