Learn More
Medchemexpress LLC Glycidyl oleate-d5 | 1426395-63-2 | MFCD32201829 | 98.8% | 343.56 | C21H33D5O3 | 10 MG
SDP
Catalog No. 5000287622
Encompass_Preferred
Glycidyl oleate-d5 is a deuterium-labeled analogue of glycidyl oleate provided for research use as an isotopic tracer and internal standard. Supplied as a liquid with high isotopic and chemical purity, it is intended for use in quantitative analytical workflows where accurate compound tracking and quantitation are required.
  • Deuterium-labeled internal standard for quantitative assays.
  • Suitable for NMR, GC-MS, and LC-MS analysis.
  • High chemical and isotopic purity for accurate quantitation.
  • Colorless to light yellow liquid appearance for easy handling.
  • Available in small pack sizes for trace-level standards.
  • Stable when stored under recommended conditions to preserve integrity.
Catalog No. 50-002-87622 Supplier Medchemexpress LLC Supplier No. HY151868S10MG
Glycidyl oleate-d5 is a deuterium-labeled analogue of glycidyl oleate provided for research use as an isotopic tracer and internal standard. Supplied as a liquid with high isotopic and chemical purity, it is intended for use in quantitative analytical workflows where accurate compound tracking and quantitation are required.
  • Deuterium-labeled internal standard for quantitative assays.
  • Suitable for NMR, GC-MS, and LC-MS analysis.
  • High chemical and isotopic purity for accurate quantitation.
  • Colorless to light yellow liquid appearance for easy handling.
  • Available in small pack sizes for trace-level standards.
  • Stable when stored under recommended conditions to preserve integrity.