Learn More
Medchemexpress LLC Glycidyl oleate (oleic acid glycidyl ester) | 5431-33-4 | MFCD01710991 | >99.0% | 338.52 | C21H38O3 | 10 MG
SDP
Catalog No. 5000284290
Encompass_Preferred
Glycidyl oleate (oleic acid glycidyl ester) is an epoxide-containing oleate ester used as a reagent in organic synthesis and lipid modification studies. Supplied for laboratory research use, it is provided in small, accurately measured quantities suitable for analytical and preparative experiments.
  • Epoxide-functional oleate ester useful for synthesis and modification of lipids
  • High reported purity (>99%) suitable for analytical and preparative work
  • Supplied in small, accurately weighed quantities for lab-scale use
  • Identifiers: CAS 5431-33-4, molecular weight 338.52
  • For research use only; not for human or clinical applications
Catalog No. 50-002-84290 Supplier Medchemexpress LLC Supplier No. HYW58593510MG
Glycidyl oleate (oleic acid glycidyl ester) is an epoxide-containing oleate ester used as a reagent in organic synthesis and lipid modification studies. Supplied for laboratory research use, it is provided in small, accurately measured quantities suitable for analytical and preparative experiments.
  • Epoxide-functional oleate ester useful for synthesis and modification of lipids
  • High reported purity (>99%) suitable for analytical and preparative work
  • Supplied in small, accurately weighed quantities for lab-scale use
  • Identifiers: CAS 5431-33-4, molecular weight 338.52
  • For research use only; not for human or clinical applications