Learn More
Cayman Chemical Glycidyl Palmitate, Purity: ≥98%, CAS: 7501-44-2, 10 mg
SDP
Catalog No. 502896027
Encompass_Preferred
A glycidyl fatty acid ester; has been found in palm and corn oils
Catalog No. 50-289-6027 Supplier Cayman Chemical Supplier No. 900368110 MG
A glycidyl fatty acid ester; has been found in palm and corn oils