Learn More
Cayman Chemical Glycidyl Palmitate, Purity: ≥98%, CAS: 7501-44-2, 50 mg
SDP
Catalog No. 502896028
Encompass_Preferred
A glycidyl fatty acid ester; has been found in palm and corn oils
Catalog No. 50-289-6028 Supplier Cayman Chemical Supplier No. 900368150 MG
A glycidyl fatty acid ester; has been found in palm and corn oils