Learn More
Medchemexpress LLC Glycidyl palmitate | 7501-44-2 | MFCD14636325 | 98.0% | 312.49 g·mol⁻1 | C19H36O3 | 10 MG
SDP
Catalog No. 5000283526
Encompass_Preferred
Glycidyl palmitate is an organic epoxide ester supplied as a high-purity research reagent for biochemical and analytical work. It is intended for use as an analytical reference, in assay development, and in synthetic applications requiring an epoxidized fatty acid ester.
  • High purity (98.0%) suitable for research-grade applications.
  • Small packaging (10 mg) convenient for analytical and assay use.
  • Epoxide functional group enables derivatization and synthetic transformations.
  • Batch-specific certificate of analysis available for traceability.
  • Supplied as a stable, well-characterized solid for reproducible results.
Catalog No. 50-002-83526 Supplier Medchemexpress LLC Supplier No. HYW12757810MG
Glycidyl palmitate is an organic epoxide ester supplied as a high-purity research reagent for biochemical and analytical work. It is intended for use as an analytical reference, in assay development, and in synthetic applications requiring an epoxidized fatty acid ester.
  • High purity (98.0%) suitable for research-grade applications.
  • Small packaging (10 mg) convenient for analytical and assay use.
  • Epoxide functional group enables derivatization and synthetic transformations.
  • Batch-specific certificate of analysis available for traceability.
  • Supplied as a stable, well-characterized solid for reproducible results.