Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycidyl palmitate | 7501-44-2 | MFCD14636325 | 98.0% | 312.49 g/mol | C19H36O3 | 100 MG
SDP

Catalog No. 5000282816
Encompass_Preferred

Glycidyl palmitate is an organic biochemical reagent (oxiran-2-ylmethyl palmitate) provided for laboratory research and analytical applications. It is supplied as a high-purity solid suitable for synthesis, standards preparation, and other life-science workflows. Store and handle according to recommended conditions; not for human or clinical use.

  • Molecular formula C19H36O3.
  • Molecular weight 312.49 g/mol.
  • CAS number 7501-44-2.
  • Purity 98.0%.
  • Physical form solid (powder).
  • Supplied as 100 mg vial.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • Intended for research use only.

Catalog No. 50-002-82816 Supplier Medchemexpress LLC Supplier No. HYW127578100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Glycidyl palmitate is an organic biochemical reagent (oxiran-2-ylmethyl palmitate) provided for laboratory research and analytical applications. It is supplied as a high-purity solid suitable for synthesis, standards preparation, and other life-science workflows. Store and handle according to recommended conditions; not for human or clinical use.

  • Molecular formula C19H36O3.
  • Molecular weight 312.49 g/mol.
  • CAS number 7501-44-2.
  • Purity 98.0%.
  • Physical form solid (powder).
  • Supplied as 100 mg vial.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.