Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycidyl palmitate | 7501-44-2 | MFCD14636325 | 98.0% | 312.49 g/mol | C19H36O3 | 250 MG
SDP

Catalog No. 5000283100
Encompass_Preferred

Glycidyl palmitate is an oxirane ester of palmitic acid used as an organic reagent and reference material in chemical and life-science research. It is provided as a high-purity compound suitable for analytical, synthetic, and assay applications, and is accompanied by standard technical documents for laboratory use.

  • High purity (98.0%).
  • Chemical formula C19H36O3.
  • Molecular weight 312.49 g/mol.
  • Available in multiple package sizes, including 250 mg.
  • Includes supporting documentation, such as SDS, COA, NMR, and MS.
  • Suitable as a reference standard or reagent for analytical and synthetic workflows.

Catalog No. 50-002-83100 Supplier Medchemexpress LLC Supplier No. HYW127578250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycidyl palmitate is an oxirane ester of palmitic acid used as an organic reagent and reference material in chemical and life-science research. It is provided as a high-purity compound suitable for analytical, synthetic, and assay applications, and is accompanied by standard technical documents for laboratory use.

  • High purity (98.0%).
  • Chemical formula C19H36O3.
  • Molecular weight 312.49 g/mol.
  • Available in multiple package sizes, including 250 mg.
  • Includes supporting documentation, such as SDS, COA, NMR, and MS.
  • Suitable as a reference standard or reagent for analytical and synthetic workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.