Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycidyl palmitate | 7501-44-2 | MFCD14636325 | 98.0% | 312.49 g/mol | C19H36O3 | 50 MG
SDP

Catalog No. 5000282682
Encompass_Preferred

Glycidyl palmitate is an epoxy fatty ester supplied as a white to off-white solid for life-science research. It serves as a biochemical reagent and intermediate for analytical and synthetic applications, with defined storage guidelines to maintain stability.

  • Epoxy ester useful for analytical and synthetic chemistry.
  • High purity: 98.0%.
  • Solid appearance: white to off-white.
  • Molecular weight: 312.49 g/mol.
  • Storage recommendations: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Available in small pack sizes, including 50 MG.

Catalog No. 50-002-82682 Supplier Medchemexpress LLC Supplier No. HYW12757850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycidyl palmitate is an epoxy fatty ester supplied as a white to off-white solid for life-science research. It serves as a biochemical reagent and intermediate for analytical and synthetic applications, with defined storage guidelines to maintain stability.

  • Epoxy ester useful for analytical and synthetic chemistry.
  • High purity: 98.0%.
  • Solid appearance: white to off-white.
  • Molecular weight: 312.49 g/mol.
  • Storage recommendations: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Available in small pack sizes, including 50 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.