Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycidyl palmitate | 7501-44-2 | MFCD14636325 | 98.0% | 312.49 g/mol | C19H36O3 | 500 MG
SDP

Catalog No. 5000282681
Encompass_Preferred

Glycidyl palmitate is the glycidyl ester of palmitic acid provided as a high-purity solid reagent for research and analytical applications. It is suitable as an analytical standard, synthetic intermediate, or biochemical reagent in life-science research, and is supplied with supporting documentation for quality control and safe handling.

  • High purity (98.0%) suitable for analytical use.
  • Solid physical form for easy handling and weighing.
  • Molecular weight 312.49 g/mol and formula C19H36O3.
  • Available in multiple pack sizes, including 500 mg.
  • Supplied with COA, data sheet, and SDS for traceability.
  • Recommended storage conditions prolong stability (powder: -20°C or 4°C).

Catalog No. 50-002-82681 Supplier Medchemexpress LLC Supplier No. HYW127578500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycidyl palmitate is the glycidyl ester of palmitic acid provided as a high-purity solid reagent for research and analytical applications. It is suitable as an analytical standard, synthetic intermediate, or biochemical reagent in life-science research, and is supplied with supporting documentation for quality control and safe handling.

  • High purity (98.0%) suitable for analytical use.
  • Solid physical form for easy handling and weighing.
  • Molecular weight 312.49 g/mol and formula C19H36O3.
  • Available in multiple pack sizes, including 500 mg.
  • Supplied with COA, data sheet, and SDS for traceability.
  • Recommended storage conditions prolong stability (powder: -20°C or 4°C).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.