Learn More
Medchemexpress LLC Glycidyl palmitate-d5 | 1794941-80-2 | >95.0% | 317.52 g/mol | C19H31D5O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW127578S10MG
Encompass_Preferred
Glycidyl palmitate-d5 is the deuterium-labeled analogue of glycidyl palmitate used as a tracer and internal standard for quantitative analytical methods. Supplied in small pack sizes, it supports calibration, method development, and accurate quantitation in laboratory assays.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS workflows.
  • Provided in small pack sizes for analytical reference use.
  • Documentation available: data sheet and handling instructions; certificate of analysis available on request.
  • Molecular formula C19H31D5O3; molecular weight 317.52 g/mol.
Catalog No. 50-002-83590