Learn More
Medchemexpress LLC Glycidyl palmitate-d5 | 1794941-80-2 | MFCD29036799 | 317.52 g/mol | C19H31D5O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW127578S5MG
Encompass_Preferred
Glycidyl palmitate-d5 is the deuterium-labeled analogue of glycidyl palmitate provided for analytical and research use. It is intended for use as a tracer and as an internal standard to support accurate quantitation in spectroscopy and mass spectrometry methods. Documentation and a certificate of analysis are available to support analytical workflows.
  • Deuterium-labeled internal standard for quantitative analysis by NMR, GC-MS, and LC-MS.
  • Suitable as a tracer for metabolic and analytical studies.
  • Provided in small, pre-weighed quantities for method development and calibration.
  • Documentation and certificate of analysis available for lot verification.
  • Molecular formula C19H31D5O3; molecular weight 317.52 g/mol.
Catalog No. 50-002-83523