Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycidyl Propargyl Ether 95.0+%, TCI America™
SDP

Catalog No. G04455G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G04455G 5 g
G04451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G04455G Supplier TCI America Supplier No. G04455G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18180-30-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00099300
InChI Key SYFZCLMMUNCHNH-UHFFFAOYSA-N
Synonym Glycidyl 2-Propynyl Ether, 2,3-Epoxypropyl Propargyl Ether
PubChem CID 281888
IUPAC Name 2-(prop-2-ynoxymethyl)oxirane
SMILES C#CCOCC1CO1

Specifications

Color Yellow
Boiling Point 95°C
Quantity 5 g
Formula Weight 112.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Glycidyl Propargyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.