Learn More
Medchemexpress LLC Glycidyl stearate-d5 | 1346598-19-3 | 345.57 | 10 MG
SDP
Catalog No. 5000451809
Encompass_Preferred
Glycidyl stearate-d5 is a deuterium labeled Glycidyl stearate. Oxiran-2-ylmethyl stearate is an ester containing reactive oxirane or epoxy groups. It is derived from stearic acid and epichlorohydrin, which are naturally occurring substances. Glycidyl stearate is commonly used in various industrial applications such as the production of coatings, adhesives, and surfactants. It can also be used as a crosslinking agent in the manufacture of polymers and resins.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-51809 Supplier Medchemexpress LLC Supplier No. HYW011188S10MG
Glycidyl stearate-d5 is a deuterium labeled Glycidyl stearate. Oxiran-2-ylmethyl stearate is an ester containing reactive oxirane or epoxy groups. It is derived from stearic acid and epichlorohydrin, which are naturally occurring substances. Glycidyl stearate is commonly used in various industrial applications such as the production of coatings, adhesives, and surfactants. It can also be used as a crosslinking agent in the manufacture of polymers and resins.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.