Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Glycidyltrimethylammonium Chloride

Catalog No. 5005350ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
2 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-053-50ML 50 mL
50-053-2L 2 L
50-053-250ML 250 mL
50-053-1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 50-053-50ML Supplier Sigma Aldrich Supplier No. 5005350ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Glycidyltrimethylammonium chloride (GTMAC) is a cationizing agent commonly used for starch modification.Glycidyltrimethylammonium chloride be used as a derivatizing agent to synthesize N-(2-hydroxypropyl)-3-trimethylammonium chitosan chloride (HTCC), a water-soluble chitosan derivative, by reacting with chitosan. HTCC can form nanocomposite films with silver nanoparticles that show good antimicrobial property and optical transparency. Cationic starches can also be prepared by derivatization of starch with GTMAC.

Specifications

CAS 3033-77-0
Density 1.13 g/mL at 20°C
Molecular Formula C6H14ClNO
Linear Formula C6H14ClNO
MDL Number MFCD00011940
Quantity 50 mL
Synonym (2,3-Epoxypropyl)trimethylammonium chloride
Molecular Weight (g/mol) 151.63 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.