Learn More
Cayman Chemical Glycine-13C, Purity: ≥95%, CAS: 20220-62-6, 100 mg
SDP
Catalog No. 502911068
Encompass_Preferred
An internal standard for the quantification of glycine by GC- or LC-MS
Catalog No. 50-291-1068 Supplier Cayman Chemical Supplier No. 35130100 MG
An internal standard for the quantification of glycine by GC- or LC-MS