Learn More
Medchemexpress LLC Glycinamide, glycylglycyl-L-phenylalanyl-N-[(carboxymethoxy)methyl]- | 1599440-20-6 | 100.0% | C18H25N5O7 | 1 G
SDP
Catalog No. 5000357884
Encompass_Preferred
Gly-Gly-Phe-Gly-NH-CH2-O-CH2COOH is an ADC linker designed for synthesizing drug-linker conjugates used in Antibody-Drug Conjugates (ADCs). It is particularly noted for its application in the synthesis of Deruxtecan, a toxic drug-linker conjugate. This product is supplied as a white to off-white solid and is intended for research use only.
  • ADC linker
  • Used to synthesize drug-linker conjugates for ADCs
  • Suitable for Deruxtecan synthesis
  • High purity
  • For research use only
Catalog No. 50-003-57884 Supplier Medchemexpress LLC Supplier No. HY456721G
Gly-Gly-Phe-Gly-NH-CH2-O-CH2COOH is an ADC linker designed for synthesizing drug-linker conjugates used in Antibody-Drug Conjugates (ADCs). It is particularly noted for its application in the synthesis of Deruxtecan, a toxic drug-linker conjugate. This product is supplied as a white to off-white solid and is intended for research use only.
  • ADC linker
  • Used to synthesize drug-linker conjugates for ADCs
  • Suitable for Deruxtecan synthesis
  • High purity
  • For research use only