Learn More
GENOVIS INC GlycINATOR, 2000 Units
Catalog No. NC0820180
Encompass
GlycINATOR (EndoS2) is an IgG-specific endoglycosidase that hydrolyzes all glycoforms present at the Fc-glycosylation site. The enzyme acts on the chitobiose core and leaves the core GlcNAc intact.
This item is not returnable. View return policy
Catalog No. NC0820180 Supplier GENOVIS INC Supplier No. A0GL1020
GlycINATOR (EndoS2) is an IgG-specific endoglycosidase that hydrolyzes all glycoforms present at the Fc-glycosylation site. The enzyme acts on the chitobiose core and leaves the core GlcNAc intact.