Learn More
Medchemexpress LLC Glycine-1-13C,15N | 112898-03-0 | MFCD00064425 | 99.5% | 77.05 g·mol⁻¹ | C13CH515NO2 | 25mg
SDP
Catalog No. 5000269784
Encompass_Preferred
Glycine-1-13C 15N is the 13C- and 15N-labeled Glycine Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors
Catalog No. 50-002-69784 Supplier Medchemexpress LLC Supplier No. HYY0966S525MG
Glycine-1-13C 15N is the 13C- and 15N-labeled Glycine Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors