Learn More
Medchemexpress LLC Glycine-1-13C,15N | 112898-03-0 | MFCD00064425 | 99.5% | 77.053 g/mol | C2H5NO2 | 5mg
SDP
Catalog No. 5000268040
Encompass_Preferred
Glycine-1-13C 15N is the 13C- and 15N-labeled Glycine Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors
Catalog No. 50-002-68040 Supplier Medchemexpress LLC Supplier No. HYY0966S55MG
Glycine-1-13C 15N is the 13C- and 15N-labeled Glycine Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors