Learn More
Medchemexpress LLC Glycine-1-13C | 20110-59-2 | MFCD00037548 | 99.4% | 76.06 | C13CH5NO2 | 50mg
SDP
Catalog No. 5000267826
Encompass_Preferred
Glycine-1-13C is the 13C-labeled Glycine Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors
Catalog No. 50-002-67826 Supplier Medchemexpress LLC Supplier No. HYY0966S450MG
Glycine-1-13C is the 13C-labeled Glycine Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors