Learn More
Medchemexpress LLC Glycine-13C2,15N | 211057-02-2 | MFCD00083979 | 99.9% | 78.05 g/mol | 13C2H515NO2 | 50mg
SDP
Catalog No. 5000268044
Encompass_Preferred
Glycine-13C2 15N is the 13C- and 15N-labeled Glycine Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors
Catalog No. 50-002-68044 Supplier Medchemexpress LLC Supplier No. HYY0966S650MG
Glycine-13C2 15N is the 13C- and 15N-labeled Glycine Glycine is an inhibitory neurotransmitter in the CNS and also acts as a co-agonist along with glutamate facilitating an excitatory potential at the glutaminergic N-methyl-D-aspartic acid (NMDA) receptors